Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xkr8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498353.2 | XP_003498401.1 | 2 - 3 | 550 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |