Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab11fip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500169.2 | XP_003500217.1 | 4 - 5 | 1349 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |