Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509434.2 | XP_003509482.1 | 3 - 4 | 288 | 91 | |
GenBank mRNA | JI887666.1 | - | 3 - 4 | 284 | 91 | |
JP046502.1 | - | 3 - 4 | 284 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |