Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650662.1 | XP_007648852.1 | 5 - 6 | 851 | 74 | |
GenBank mRNA | JI874541.1 | - | 5 - 6 | 701 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Recommended Assay Probe spans exons |