Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650662.1 | XP_007648852.1 | 6 - 7 | 904 | 93 | |
GenBank mRNA | JI874541.1 | - | 6 - 7 | 754 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Probe spans exons |