Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100771726 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652260.1 | XP_007650450.1 | 18 - 19 | 2175 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |