Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654349.1 | XP_007652539.1 | 4 - 5 | 725 | 96 | |
GenBank mRNA | JP055628.1 | - | 4 - 5 | 684 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |