Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcng3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496231.1 | XP_003496279.1 | 1 - 2 | 59 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel 3' Most Recommended Assay Probe spans exons |