Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nptx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508163.1 | XP_003508211.1 | 4 - 5 | 669 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |