Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510039.1 | XP_003510087.1 | 13 - 14 | 2162 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |