Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eri2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511944.2 | XP_003511992.1 | 3 - 4 | 210 | 80 | |
XM_007652449.1 | XP_007650639.1 | 3 - 4 | 210 | 80 | ||
GenBank mRNA | JP048241.1 | - | 3 - 4 | 204 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |