Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653495.1 | XP_007651685.1 | 5 - 6 | 525 | 57 | |
GenBank mRNA | JI866438.1 | - | 4 - 5 | 449 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |