Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647415.1 | XP_007645605.1 | 8 - 9 | 1045 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |