Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3il1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513179.2 | XP_003513227.1 | 8 - 9 | 1095 | 100 | |
XM_007653498.1 | XP_007651688.1 | 7 - 8 | 876 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |