Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497082.2 | XP_003497130.1 | 3 - 4 | 946 | 80 | |
XM_007607713.1 | XP_007605903.1 | 2 - 3 | 873 | 80 | ||
XM_007607717.1 | XP_007605907.1 | 3 - 4 | 946 | 80 | ||
GenBank mRNA | JI876561.1 | - | 3 - 4 | 911 | 80 | |
JP064842.1 | - | 3 - 4 | 911 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |