Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hlf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498185.1 | XP_003498233.1 | 3 - 4 | 669 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |