Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508442.2 | XP_003508490.1 | 2 - 3 | 152 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |