Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spocd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007627072.1 | XP_007625262.1 | 2 - 3 | 827 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Probe spans exons |