Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497665.2 | XP_003497713.1 | 4 - 5 | 1307 | 66 | |
XM_007615556.1 | XP_007613746.1 | 3 - 4 | 1412 | 66 | ||
XM_007615568.1 | XP_007613758.1 | 3 - 4 | 1415 | 66 | ||
XM_007615577.1 | XP_007613767.1 | 3 - 4 | 1083 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |