Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003506917.2 | XP_003506965.1 | 2 - 3 | 1735 | 71 |