Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646904.1 | XP_007645094.1 | 2 - 3 | 158 | 77 | |
XM_007646905.1 | XP_007645095.1 | 2 - 3 | 158 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |