Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495141.2 | XP_003495189.1 | 2 - 3 | 1003 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |