Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497511.2 | XP_003497559.2 | 1 - 2 | 966 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |