Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500902.2 | XP_003500950.1 | 1 - 2 | 285 | 65 | |
GenBank mRNA | JI891025.1 | - | 1 - 2 | 285 | 65 | |
JP057317.1 | - | 1 - 2 | 285 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |