Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h12c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643376.1 | XP_007641566.1 | 4 - 5 | 1466 | 75 | |
GenBank mRNA | JI876926.1 | - | 4 - 5 | 674 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |