Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505142.2 | XP_003505190.1 | 9 - 10 | 3023 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |