Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644151.1 | XP_007642341.1 | 8 - 9 | 1169 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Probe spans exons |