Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505470.1 | XP_003505518.1 | 7 - 8 | 946 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |