Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neurod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508936.1 | XP_003508984.1 | 1 | 524 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Both primers and probe map within a single exon |