Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rxra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653390.1 | XP_007651580.1 | 5 - 6 | 895 | 86 | |
GenBank mRNA | JI879460.1 | - | 1 - 2 | 113 | 86 | |
JP049010.1 | - | 1 - 2 | 113 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Nuclear Receptor Probe spans exons |