Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nppb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514161.2 | XP_003514209.1 | 2 - 3 | 435 | 67 | |
GenBank mRNA | JP053352.1 | - | 1 - 2 | 134 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |