Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mthfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514165.2 | XP_003514213.1 | 10 - 11 | 2144 | 85 | |
XM_007654352.1 | XP_007652542.1 | 10 - 11 | 2141 | 85 | ||
XM_007654353.1 | XP_007652543.1 | 9 - 10 | 1775 | 85 | ||
XM_007654354.1 | XP_007652544.1 | 10 - 11 | 1800 | 85 | ||
XM_007654355.1 | XP_007652545.1 | 10 - 11 | 1797 | 85 | ||
XM_007654356.1 | XP_007652546.1 | 10 - 11 | 1663 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |