Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651581.1 | XP_007649771.1 | 7 - 8 | 739 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |