Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc9c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514301.2 | XP_003514349.1 | 1 - 2 | 579 | 70 | |
XM_007654455.1 | XP_007652645.1 | 2 - 3 | 1040 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |