Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502413.1 | XP_003502461.1 | 3 - 4 | 1775 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel 3' Most Probe spans exons |