Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rgs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502854.1 | XP_003502902.1 | 4 - 5 | 362 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |