Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam86a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508119.2 | XP_003508167.1 | 1 - 2 | 116 | 55 | |
GenBank mRNA | JI888750.1 | - | 1 - 2 | 118 | 55 | |
JP050303.1 | - | 1 - 2 | 118 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |