Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511175.1 | XP_003511223.1 | 6 - 7 | 1396 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase 3' Most Probe spans exons |