Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642919.1 | XP_007641109.1 | 7 - 8 | 758 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Probe spans exons |