Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501951.2 | XP_003501999.1 | 1 - 2 | 169 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Recommended Assay Probe spans exons |