Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514790.2 | XP_003514838.1 | 4 - 5 | 1929 | 75 | |
GenBank mRNA | JI869466.1 | - | 1 - 2 | 497 | 75 | |
JP047205.1 | - | 1 - 2 | 497 | 75 |