Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650314.1 | XP_007648504.1 | 5 - 6 | 2083 | 99 | |
XM_007650315.1 | XP_007648505.1 | 3 - 4 | 1865 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Probe spans exons |