Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sycp2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647029.1 | XP_007645219.1 | 15 - 16 | 1449 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |