Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640914.1 | XP_007639104.1 | 8 - 9 | 1660 | 75 | |
XM_007640915.1 | XP_007639105.1 | 7 - 8 | 505 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Probe spans exons |