Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509537.2 | XP_003509585.1 | 1 - 2 | 214 | 71 | |
GenBank mRNA | JI879283.1 | - | 1 - 2 | 47 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Probe spans exons |