Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Narg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003513659.2 | XP_003513707.1 | 15 - 16 | 3017 | 55 | |
XM_007653918.1 | XP_007652108.1 | 15 - 16 | 2947 | 55 | ||
XM_007653919.1 | XP_007652109.1 | 15 - 16 | 3114 | 55 | ||
XM_007653920.1 | XP_007652110.1 | 12 - 13 | 2435 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Probe spans exons |