Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frem3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007646784.1 | XP_007644974.1 | 7 - 8 | 5765 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Probe spans exons |