Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503712.2 | XP_003503760.1 | 2 - 3 | 1929 | 72 | |
XM_007644287.1 | XP_007642477.1 | 2 - 3 | 1929 | 72 | ||
XM_007644288.1 | XP_007642478.1 | 3 - 4 | 1806 | 72 | ||
XM_007644289.1 | XP_007642479.1 | 2 - 3 | 1726 | 72 | ||
XM_007644290.1 | XP_007642480.1 | 2 - 3 | 1065 | 72 | ||
XM_007644291.1 | XP_007642481.1 | 2 - 3 | 825 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |