Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mei4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510873.2 | XP_003510921.2 | 3 - 4 | 844 | 94 | |
XM_007651459.1 | XP_007649649.1 | 3 - 4 | 833 | 94 |