Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007652106.1 | XP_007650296.1 | 6 - 7 | 875 | 62 | |
XM_007652145.1 | XP_007650335.1 | 6 - 7 | 872 | 62 | ||
XM_007652188.1 | XP_007650378.1 | 6 - 7 | 845 | 62 | ||
GenBank mRNA | JP053164.1 | - | 6 - 7 | 900 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |